Flasker, bred hals, med gevind
Leverandør: KJS KUNSTSTOFFVERPACKUNGEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
W50 W100 W250 W500 W1000 W2000
215-6001EA 14 DKK
215-6001 215-6002 215-6003 215-6004 215-6005 215-6007
Flasker, bred hals, med gevind
Flasker Plastflasker
LDPE, naturlig farve.
  • Good chemical resistance
  • Leakproof, tough and versatile
Perfect for sample collection, storage and transport. Ideal for liquids, paste-like media and granular material. The wide mouth allows easy filling and cleaning. Without graduation.
Bestillingsoplysninger: Please order screw caps separately.