Bottles, wide neck, with screw cap, Azlon®
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BWP0250PGRAD BWP0500PGRAD BWP1000PGRAD BWP0060PGRAD
215-0555EA 496 DKK
215-0555 215-0557 215-0559 215-0092
Bottles, wide neck, with screw cap, Azlon®
Flasker Plastflasker
PP, high clarity, with PP screw cap.
  • Leakproof, liner-less design
  • Clear crisp printed graduations show millilitres and fluid ounces
  • Printed with large white 'write on' panel for easy labelling
  • Autoclavable at 121 °C
  • Smooth inner contour for efficient pouring and ease of cleaning
Ideal for both liquids and solids, perfect for microbiological sampling.
When autoclaving bottles always ensure the caps are completely loosened or removed to prevent accidental implosion.