Flasker, fluorideret, bred hals, med skruelåg, Nalgene®
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2197-0004 2197-0008 2197-0016 2197-0032
215-1039EA 410 DKK
215-1039 215-1040 215-1041 215-1042
Flasker, fluorideret, bred hals, med skruelåg, Nalgene®
Flasker Plastflasker
HDPE, fluorideret, med fluorideret PP-skruelåg.
  • Safe, durable, fluorinated HDPE bottle has fluorocarbon surface, inside and out
  • Fluorinated surface enhances long-term performance and prevents permeation loss
  • Leakproof
Fluorinating these HDPE bottles provides a cost effective alternative to costly fluoropolymer containers. Wide mouth makes filling and emptying of solid and liquid substances easier. Crack and impact resistant.