Weighing bottles, glass
Leverandør: Lenz Laborglas GmbH & CO.KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
05565334 05565340 05565350 05565380 05565550 05565734 05565740 05565750 05565540 05565534 05565580 05565780
611-1236EA 223 DKK
611-1236 611-1237 611-1238 611-1239 611-9308 611-9321 611-9320 611-9319 611-9323 611-9324 611-9325 611-9318
Weighing bottles, glass
Flasker Vejeflasker
DURAN®, borosilicate glass.
  • With interchangeable cap
  • Exterior joint improves material transfers and reduces risk of material contamination of cap
  • Available in low, medium or tall forms