Wash bottles, narrow neck
Leverandør: MURRAY S
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
L780/01 L780/02
215-0965EA 122 DKK
215-0965 215-0966
Wash bottles, narrow neck
Flasker Sprøjteflasker
LDPE, translucent with jet.
  • Good chemical resistance
  • Heat resistant up to +80 °C
  • Leakproof
Economical, soft and very easy to press. For use with general solutions such as water, soap, dilute acids and dilute alkalis.