Bottles, narrow neck, with screw cap, Bijou type
Leverandør: MURRAY S
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
G751/01 G751/11 G752/01
215-3147EA 2930 DKK
215-3147 MURRG751/11 215-3146
Bottles, narrow neck, with screw cap, Bijou type
Flasker Glasflasker
Flint glass, clear with aluminium screw caps.
  • Bijou, media or universal type
  • With fitted aluminium rubber lined cap
  • Sulphated for autoclaving