Bottles, narrow neck, with standard ground stoppers
Leverandør: GLASWARENFABRIK KARL HECHT
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
41280006 41280012 41280018 41280024 41280030 41280036 41282006 41282012 41282018 41282024 41282030 41282036
215-1253EA 69.3 DKK
215-1253 215-1254 215-1255 215-1256 215-1257 215-1258 215-1273 215-1274 215-1275 215-1276 215-1277 215-1278
Bottles, narrow neck, with standard ground stoppers
Flasker Glasflasker
Soda-lime glass, clear or amber, with standard ground joint and HDPE stopper.
  • Good chemical and physical properties
  • For use in applications that are not associated with high thermal loads
  • Stable and robust
Amber coloured to protect light-sensitive materials.
DIN 12036, ISO 4796
Certificeringer: DIN 12036, ISO 4796.