Flasker, smal hals, LDPE, med gevind
Leverandør: KJS KUNSTSTOFFVERPACKUNGEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
E50 E100 E250 E500 E1000
215-6008EA 14 DKK
215-6008 215-6009 215-6010 215-6011 215-6012
Flasker, smal hals, LDPE, med gevind
Flasker Plastflasker
LDPE, natural colour.
  • Squeezable, so these flasks make excellent containers for droppers or wash bottles/sprayers
  • Please order caps (screw, dropper or spray caps) separately
Excellent for sampling, storing and transporting liquids.