Infusion bottles
Leverandør: NORDIC PACK
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6991 6781 6551 36-0 3041
215-3769EA 88.6 DKK
215-3769 215-3768 215-3767 215-3764 215-3763
Infusion bottles
Flasker Infusionsflasker
Glass, clear.
  • Good chemical resistance
  • Good temperature resistance
  • Screw caps must be ordered seperately
Infusion bottles with thread.