Bottles, narrow neck, square, technical treatments, with thread
Leverandør: MELLERUD PLAST EMBALLATOR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
48E0100320 48E0015220 48E0010220 48E0025220 48E0050320
215-3074EA 33.9 DKK
215-3074 215-3068 215-3067 215-3069 215-3071
Bottles, narrow neck, square, technical treatments, with thread
Flasker Plastflasker
HDPE, natural, without screw cap.
  • Square, space-saving
  • Screw cap must be ordered separately
For packaging, sample collection and storage.