Bottles, rectangular, narrow neck with screw caps, Corning® Gosselin™
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
FV250-05 FV250-09 FV500-04
215-0700EA 3990 DKK
215-0700 215-0699 215-0702
Bottles, rectangular, narrow neck with screw caps, Corning® Gosselin™
Flasker Plastflasker
HDPE, translucent, graduated with red tamper evident screw caps.
  • Rectangular cross section bottles save storage space
  • Graduated bottles are supplied with caps but are unassembled
  • Sterile bottles are irradiated and are supplied capped
These bottles are graduated and sterile available. Ideal for use in laboratory, production plant or in the field for storage of liquids, solids and powders.