Flasker, smal hals, rund, HDPE, med skruelåg, Nalgene®
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2097-0005 2097-0008 2097-0010 2097-0016 2097-0020 2097-0032 2097-0050
216-8504EA 275 DKK
216-8504 216-8501 216-8505 216-8502 216-8506 216-8503 216-8507
Flasker, smal hals, rund, HDPE, med skruelåg, Nalgene®
Flasker Plastflasker
HDPE, fluorideret, med fluorideret PP-skruelåg.
  • Fluorinated bottles and carboys can be used for most acids, alkalis and aggressive organic solvents
  • Heavy-duty walls are safe, durable and resistant to splitting and punctures
  • Fluorocarbon surfaces inside and out
  • Leakproof
Fluorinating these HDPE bottles and carboys provides a cost effective alternative to costly fluoropolymer containers.