Emballageflasker, smal hals, med gevind, Nalgene®
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
362089-0001 362089-0004 362089-0002 362089-0008 362089-0016 362089-0032
215-1234EA 4640 DKK
215-1234 215-1235 215-1587 215-1588 215-1589 215-1590
Emballageflasker, smal hals, med gevind, Nalgene®
Flasker Plastflasker
HDPE, translucent.
  • Good chemical resistance
  • Bottles and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI
  • Supplied without closures; leakproof sealing with closures 215-1597/215-1252/215-1598/215-1611/215-1236/215-1247/215-1613
Economical and robust bottles for storing and transporting liquids.