Flasker, smal hals, rund, HDPE, lavt partikelindhold, IP2, med skruelåg, Nalgene®
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
382099-0250 382099-0500 382099-1000
215-1271EA 372 DKK
215-1271 215-1272 215-1279
Flasker, smal hals, rund, HDPE, lavt partikelindhold, IP2, med skruelåg, Nalgene®
Flasker Plastflasker
HDPE, gennemskinnelig med PP skruelåg.
  • Good chemical resistance
  • Screw cap supplied assembled on the bottle
  • Leakproof
Bottles with an average of less than 30 particles per ml at 0,3 µm and greater. Excellent for storing high-purity chemicals. Product includes a certificate of quality that assures the product has been tested and accepted in accordance with Nalgene® specifications. Low particulate IP 2 bottles can be used in compliance with the regulations for customers who are designing, assembling and certifying their own combination packaging.