Flasker, smal hals, rund, FEP, med skruelåg, Nalgene®
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1600-0004 1600-0008 1600-0016 1600-0032 1600-0064 1600-0002 1600-0001
215-7123EA 740 DKK
215-7123 215-7124 215-7125 215-7126 215-7127 215-3695 215-7121
Flasker, smal hals, rund, FEP, med skruelåg, Nalgene®
Flasker Plastflasker
FEP, translucent, with ETFE screw cap.
  • Bedst mulige kemiske resistens
  • Temperaturresistent fra −105 °C til +150 °C, lækagesikret
  • Autoklaverbare
Bottles with the highest chemical and corrosion resistance. Suitable for applications at low and high temperatures, trace analyses, and for many types of work with organic solvents. Suitable for use in cleanrooms, analysis and for storage.