Packaging bottles, narrow neck, round, HDPE, with screw caps, Nalgene®
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
312085-0001 312089-0001 312089-0004 312089-0032 312085-0002 312085-0004 312085-0008 312085-0016 312085-0032 312089-0002 312089-0008 312089-0016
215-1007EA 8370 DKK
215-1007 215-1009 215-1017 215-1432 215-1410 215-1411 215-1412 215-1414 215-1415 215-1416 215-1430 215-1431
Packaging bottles, narrow neck, round, HDPE, with screw caps, Nalgene®
Flasker Plastflasker
HDPE, translucent or amber with PP screw cap.
  • Good chemical resistance
  • Heat resistant from –100 to +120 °C
  • Leakproof
Economical, robust bottles for storing and transporting powders, liquids or pastes. Amber bottles to protect light-sensitive substances.
Advarsel: The product(s) on this page are outside the scope of the EU Directive 2019/904.