Jars, low form, with Twist-off thread
Leverandør: RIXIUS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1-0800-0050-43 1-0800038066KT 1-0800-0720-66 1-0800-0212-66 1-0800-0720-82 1-0801-0490-82 1-0800-0160-48 1-0800-0370-66 1-0800-1062-82 S080001004875
216-3620EA 10.8 DKK
216-3620 216-3622 216-3623 216-3621 RIXI1-0800-0720-82 RIXI1-0801-0490-82 RIXI1-0800-0160-48 RIXI1-0800-0370-66 RIXI1-0800-1062-82 RIXIS080001004875
Jars, low form, with Twist-off thread
Dåser Dåser, allround
Glass, without cap.
  • Good chemical and physical properties
  • For use in applications without high thermal loads
  • Fitting metal screw caps are available under codes 216-3630 to 216-3633
Perfect for sample collection, storing and transporting of powders, granulates, pastes or liquids.