Bottle heads for gas wash bottles, Drechsel Pattern, Quickfit®
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MF28/3/125/SC MF28/3/250/SC MF28/3/500/SC
201-0774EA 565 DKK
201-0774 201-0775 201-0776
Bottle heads for gas wash bottles, Drechsel Pattern, Quickfit®
Låg Flaskelåg
PYREX®, borosilicate glass, without glass filter disc.
  • Meget god kemisk resistens
  • Høj temperaturresistens
With detachable plastic screwthreads connectors for easy fitting of flexible tubing.
Certificeringer: BS 2461
Bestillingsoplysninger: Drechsel bottle to be ordered separately.