Gas washing bottles, Drechsel pattern, VitraPOR®
Leverandør: ROBU GLASFILTER
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
41101 41100 41102 41103 41104
201-1901EA 752 DKK
201-1901 201-0922 201-0923 201-0924 201-0925
Gas washing bottles, Drechsel pattern, VitraPOR®
Flasker Gasvaskeflasker
DURAN®, borosilicate glass, clear, with PBT screw caps.
  • Sintered glass filter porosity 1 (sinter/disc diameter 25 mm)
  • All system components can be ordered separately
  • Excellent chemical resistance and temperature resistance
Comprises of gas washing bottles with screw thread DIN GL 45, versatile use - screw cap and 2 tube connectors made from PP with threaded connection.