Gasvaskeflasker, Drechsel-mønster
Leverandør: Lenz Laborglas GmbH & CO.KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
05500537 05500549 05500558 05500570 05510137 05510149 05510158 05510170 05510237 05510037 05510049 05510058 05510070 05510249 05510258 05510270
201-1421EA 433 DKK
201-1421 201-1422 201-1423 533-2124 201-1111 201-1420 201-1413 201-1414 LENZ05510237 LENZ05510037 LENZ05510049 LENZ05510058 LENZ05510070 LENZ05510249 LENZ05510258 LENZ05510270
Gasvaskeflasker, Drechsel-mønster
Flasker Gasvaskeflasker
DURAN®, borosilicate glass.
  • Available with or without sintered glass disc, porosity grade 0/1/2
  • All system components can be ordered separately
  • Excellent chemical and temperature resistance
Consists of the gas washing bottle with NS 29/32 standard ground joint socket and the insert with NS 29/32 cone joint and two glass tubing connectors with 11 mm external diameter.
Certificeringer: DIN 12463 (Gas washing bottle, bottle only); DIN 12596 (Gas washing bottle heads).