Gas washing bottles
Leverandør: Lenz Laborglas GmbH & CO.KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
05530058 05530158
201-1197EA 545 DKK
201-1197 201-1198
Gas washing bottles
Flasker Gasvaskeflasker
In Duran® glass, transparent, with PBT screw caps.
  • With or without porosity 1 filter plate (cup diameter 25 mm)
  • The components are available separately
  • Very good chemical and heat resistance
Wash bottle with DIN GL 45 screw pitch and adjustable trim with screw cap and 2 PP knobs with bolting.