Spray bottles
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
144190 144180
215-0110EA 657 DKK
215-0110 215-2320
Spray bottles
Flasker Sprayflasker
HDPE, hvid.
  • Good chemical resistance
  • Good temperature resistance
  • Robust, convenient, stable
By turning the nozzle, delivery can be varied from a fine spray to a solid stream.