Weighing boats, PYREX®
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2115/04M 2115/06M 2115/02M
611-0010EA 281 DKK
611-0010 611-0011 611-3138
Weighing boats, PYREX®
Vejebåde Vejebåde
Borosilicate glass.
  • Ideal for weighing small quantities of powder
  • With hollow stem to facilitate introduction of the powder into narrow neck vessels e.g. volumetric flasks
  • Flat base ensures stability on the balance pan