Målecylindere, konisk, Nalgene®
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3673-0004 3673-0008 3673-0016 3673-0032
NALG3673-0004EA 228 DKK
NALG3673-0004 213-0259 213-0260 213-0261
Målecylindere, konisk, Nalgene®
Cylindere Målecylindre
PMP, transparent.
  • Excellent chemical resistance
  • Unbreakable, autoclavable
  • The liquid volume can be read easily because no meniscus is formed
Ideal for pharmaceutical applications. Graduations in ml and oz. With octagonal foot for improved stability, and spout.
Certificeringer: Conforms to the volume and accuracy requirements of ASTM E1094.