VWR®, Målebægre, med håndtag
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
213-3401EA 516 DKK
213-3401 213-3402 213-3403 213-3404
VWR®, Målebægre, med håndtag
Bægre
PP, highly translucent.
  • Good chemical resistance
  • Resistant to heat up to +121 °C (for 20 minutes), prolonged use +100 °C
  • Smooth surfaces for easy, fast cleaning
  • With embossed scale, blue graduations and spout
Ideal for general laboratory use and a wide variety of other applications.
Certificeringer: Food-safe products acc. to EC Directive No. 10/2011.