Bags, autoclave, 121 °C, Sterilin®
Leverandør: STERILIN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
509-17 509L 511-17 510 510L
129-0000EA 869 DKK
129-0000 129-0001 129-0004 129-0002 129-0003
Bags, autoclave, 121 °C, Sterilin®
Poser Affaldsposer
Specifically designed for contaminated waste disposal in autoclaves or incinerators. Strong, HDPE, white, with blue 'Biohazard' printing.
  • Suitable for high temperature sterilisation at 121 °C
  • Convenient tissue box style cartons dispense bags individually
Polypropylene bags are suitable for the inactivation of particularly resistant biological waste at 135 °C. Coated wire stands or cardboard holders available for use with autoclave bags.