Autoclavable bags, transparent, Clavies®, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
H131820812 H131821010 H131852436 H131851224 H131852430 H131820810 H131820815 H131821012 H131821015
129-1017EA 1360 DKK
129-1017 129-4950 129-1021 129-1019 129-1020 129-1016 129-1018 129-1107 129-1174
Autoclavable bags, transparent, Clavies®, SP Bel-Art
Poser Affaldsposer
Autoclavable bags, in PP, 50 µm thick, transparent useful for discarding used Petri dishes, membrane filters, multi-well cell culture plates, cell culture flasks, culture plates, pipettes and more.
  • For maximum protection and convenience when disposing of biohazardous waste
  • Three largest sizes hold 65, 200 and 240 standard Petri dishes respectively
  • Steam autoclavable at 135 °C