VWR®, Poser med vridlukning
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
129-0012EA 2220 DKK
129-0012 129-0013 129-0010 129-0011 129-9870 129-9884 129-9885 129-9886 129-9887 129-0417 129-0418 300-0107 129-9019 129-9020 129-9021 129-9022 129-9023 129-9024 129-9025 129-9026 129-9027 129-9028 129-1161 129-1162 129-9800 129-9801 129-9802 129-9803 129-9804 129-9805 129-9806 129-9808 129-9807 129-9809 129-9810 129-9813 129-9814 129-9815 129-9816 129-9817 129-9029
VWR®, Poser med vridlukning
Poser Prøveposer
LDPE bags with safety tabs, manufactured and packed in sterile conditions.
  • Provide a secure, contaminant-free flexible container that ensures dependable analytical results
  • R-Nase, D-Nase, Pyrogen og BPA fri
  • Offers an economical and efficient way to collect, contain and carry samples
  • Wire tab keeps bag open for filling
  • Unique dobble fold over system provides a temporary leakproof seal
Universal use for samples such as foods, liquids, grains, petroleum products, animal material, waste and soil samples. Market segment Environmental sampling (surfaceMore Product Information