Adapters, delivery, with vent, QUICKFIT®
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
RA2/11 RA2/22
271-0008EA 458 DKK
271-0008 271-0009
Adapters, delivery, with vent, QUICKFIT®
Destillations komponenter Destillationsadaptere
PYREX®, borosilicate glass, bend.
  • Avoids the problems of 'closed systems'
With a side arm for venting purposes, not for connection to a vacuum source. For adapters with vacuum connection, see 201-8653.