Adapters, receiver, vacuum, Quickfit®
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
RA3/22/SC RA3/44/SC RA3/11/SC RA3/23/SC RA3/33/SC
201-8160EA 790 DKK
201-8160 201-0599 201-0597 BIBBRA3/23/SC 201-0598
Adapters, receiver, vacuum, Quickfit®
Destillations komponenter Destillationsadaptere
PYREX®, borosilicate glass, bent.
  • Easy connection to flexible tubing via detachable plastic screw thread connector
Adapter with a side arm for connection to a vacuum supply.