Adapters, receiver, with short or long stem, Quickfit®
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
RA1/14 RA1/03 RA1/01 RA1/02 RA1/11 RA1/12 RA1/13 RA1/04
BIBBRA1/14EA 1560 DKK
BIBBRA1/14 271-0004 271-0002 271-0003 271-0005 201-0618 201-0619 BIBBRA1/04
Adapters, receiver, with short or long stem, Quickfit®
Destillations komponenter Destillationsadaptere
PYREX®, borosilicate glass.
  • Meget god kemisk resistens
  • Høj temperaturresistens
Useful for feeding distillate from condensers into receiving vessels.