Adapters, still head, recovery, Quickfit®
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SH1/44 SH1/11 SH1/22 SH1/23 SH1/33
201-8163EA 446 DKK
201-8163 201-3046 201-3047 201-3048 BIBBSH1/33
Adapters, still head, recovery, Quickfit®
Destillations komponenter Hoveder
PYREX®, borosilicate glass.
  • Meget god kemisk resistens
  • Høj temperaturresistens
Provides a simple vapour conduit between flasks and distillation condensers.