Adapters, cone/screw thread, Quickfit®
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ST51/13 ST52/13 ST54/13 ST54/24 ST53/13 ST51/18 ST53/18 ST53/24 ST52/18 ST52/24 ST54/18
201-0219EA 629 DKK
201-0219 201-0220 201-0698 201-0699 201-0697 201-0696 201-0252 201-0227 201-0225 201-0226 201-0228
Adapters, cone/screw thread, Quickfit®
Adapter Adaptere med NS-slib
PYREX®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Screw thread joint complete with plastic cap, silicone rubber ring and PTFE washer. Ideal for securing thermometers, gas inlet tubes etc.