Sealing tapes, Nunc™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
232701 232702 276014 249719 249720 235306 235307 241205 232698 232699 236272 236370
391-7534EA 1690 DKK
391-7534 734-2134 391-7614 734-2135 NUNC249720 734-1341 732-0851 NUNC241205 391-7500 NUNC232699 NUNC236272 NUNC236370
Sealing tapes, Nunc™
Forsegling Mikrotiterpladetætninger
Effective sealing of all microplate formats. For storage, PCR, microscopy, culture and protection.
  • Pre-cut, plate-sized sheets
  • Pierceable tapes available