Beholder, firkantet, med låg, Nalgene®
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
14200-0002 14200-0005 14200-0010 14200-0015 14200-0020 14200-0045
216-1921EA 2170 DKK
216-1921 216-1922 216-1923 216-1924 216-1925 NALG14200-0045
Beholder, firkantet, med låg, Nalgene®
Tanke Firkantede beholdere
These PP rectangular tanks with cover can significantly reduce evaporation and contamination.
  • Good stress-crack resistance
  • Suitable for use with many organic chemicals
  • Tanks can be modified with a factory-installed spigot