Nalgene® Bottles, Narrow Neck, Round, PPCO, Packaging, with Screw Caps
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2087-0001 2087-0002 2087-0004 2087-0008
215-2236EA 695 DKK
215-2236 215-2237 215-2239 215-2240
Nalgene® Bottles, Narrow Neck, Round, PPCO, Packaging, with Screw Caps
Flasker Plastflasker
PPCO, translucent with natural PP screw cap.
  • Lightweight
  • Very good chemical resistance
  • Very good temperature resistance
  • Linerless closure
Perform light-duty, general purpose applications where a heavier bottle is not required.
Economical and recyclable; designed for single use. Translucence allows for easy viewing of liquid levels.
Linerless caps ensure leakproof protection without the use of liner that can wrinkle, cause leaks or contaminate the reagents.
Manufactured from non-toxic materials.
Certificeringer: Manufactured in complianceMore Product Information