2,6-Dimethyl-4-heptanol ≥85.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Diisobutylcarbinol

D0732-25ML D0732-500ML
TCIAD0732-25MLEA 132 DKK
TCIAD0732-25ML TCIAD0732-500ML
2,6-Dimethyl-4-heptanol ≥85.0% (by GC)
2,6-Dimethyl-4-heptanol
Formel: C₉H₂₀O
Molekylvægt: 144.26 g/mol
Kogepunkt: 179°C
Smeltepunkt: -65
Flammepunkt: 74°C
MDL Number: MFCD00008944
CAS-nummer: 108-82-7
EINECS: 203-619-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f99e453f608d8577a43e3891a98a868baf042f06&vid=f99e453f608d8577a43e3891a98a868baf042f06&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 85.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR