α-Hydroxy-o-tolyl β-D-glucopyranosid

Leverandør: Roth Carl
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-(Hydroxymethyl)phenyl-β-D-glucopyranoside, Salicoside, D(-)-Salicin, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(2-(hydroxymethyl)phenoxy)tetrahydro-2H-pyran-3,4,5-triol, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxane-3,4,5-triol, D(−)-Salicin

7192.1 7192.2
ROTH7192.1EA 0 DKK
ROTH7192.1 ROTH7192.2
α-Hydroxy-o-tolyl β-D-glucopyranosid
α-Hydroxy-o-tolyl β-D-glucopyranosid
Formel: C₁₃H₁₈O₇
Molekylvægt: 286.28 g/mol
Smeltepunkt: 199…201 °C
Massefylde: 1.42 g/cm³ (20 °C)
MDL Number: MFCD00006590
CAS-nummer: 138-52-3
EINECS: 205-331-6
Merck Index: 12,08476

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR