EGTA (Ethylenglycol bis(2-aminoethyl ether)-N,N,N',N'-tetraeddikesyre)
Leverandør: PanReac AppliChem
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: EGTA, Ethylenebis(oxyethylenenitrilo)tetra(acetic acid), Kestranal® FE PA, Idranal® VI, Ethylene glycol-O,O'-bis(2-aminoethyl)-N,N,N',N'-tetraacetic acid, Ethylenebis(oxyethylenenitrilo)tetraacetic acid, Ethylene Glycol Bis(2-aminoethyl Ether)-N,N,N,N-tetraacetic acid, 3,12-Bis(carboxymethyl)-6,9-dioxa-3,12-diazatetradecanedioic acid, Titriplex® VI
A0878.0025 A0878.0100
A0878.0025EA 1570 DKK
A0878.0025 A0878.0100
EGTA (Ethylenglycol bis(2-aminoethyl ether)-N,N,N',N'-tetraeddikesyre)
EGTA (Ethylenglycol bis(2-aminoethyl ether)-N,N,N',N'-tetraeddikesyre)
Formel: C₁₄H₂₄N₂O₁₀
Molekylvægt: 380.35 g/mol
Smeltepunkt: 240…244 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00004291
CAS-nummer: 67-42-5
EINECS: 200-651-2
Merck Index: 13,03560