Diaphragm laboratory pump, Sartolab® P20
Leverandør: Sartorius
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
180MP-1 180MP-1B 180MP-1F
SART180MP-1EA 15480 DKK
SART180MP-1 SART180MP-1B SART180MP-1F
Diaphragm laboratory pump, Sartolab® P20
Pumper Membranmålepumper
For the filtration of liquids with Sartolab® P20 filters.
  • Safety mechanism
  • Compact design
  • User-friendly interface
  • Dry running capability
  • Robust construction
  • Versatile control
The Sartolab® P20 Pump is a specialized fluid feed pump optimized for use with Sartolab P20 pressure filtration devices. A key safety feature of this pump is the integrated pressure relief valve, which is pre-set to a maximum of 3.5 bar relative pressure. This valve serves as an essential protective mechanism, automatically activating to relieve pressure in the event of filter blockage that results in pressure buildup, thereby maintaining a secure and reliable operation.
Dimensions: 13×99×177 mm