Millex® Automation-Compatible Filter Units, with Hydrophilic LCR (PTFE) Membrane
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SLCRDZ5NK SLCRBZ5NK SLCRBZ5NZ SLCRDZ5NZ SLLGDZ5NK SLLGDZ5NZ
516-6553EA 20440 DKK
516-6553 MILFSLCRBZ5NK MILFSLCRBZ5NZ MILFSLCRDZ5NZ MILFSLLGDZ5NK MILFSLLGDZ5NZ
Millex® Automation-Compatible Filter Units, with Hydrophilic LCR (PTFE) Membrane
Filtre Sprøjtefiltre
Automation compatible filter units are specially designed to provide efficient filtration for dissolution and other automated sample preparation applications.
  • Domed housing ensures reliable delivery of filters
  • Pressure-resistant housing resists bursting
  • Luer-Lok® connection optimised for precise alignment and fit
Automation compatible filter units are available in a variety of membrane types and pore sizes, including units with pre-filters for particulate-laden samples. Used in the process of drug dissolution testing, automated sample preparation, clarification of solutions containing high particle levels etc.
Max. temperature: 45 °C
Applications: drug dissolution testing, automated sample preparation, clarification of solutions containing high particle levels.