Stericup® Quick Release-GV Sterile Vacuum Filtration Systems
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
S2GVT05RE S2GVU05RE S2GVU11RE S2GVU02RE S2GVU01RE
MILFS2GVT05REEA 1220 DKK
MILFS2GVT05RE MILFS2GVU05RE MILFS2GVU11RE MILFS2GVU02RE MILFS2GVU01RE
Stericup® Quick Release-GV Sterile Vacuum Filtration Systems
Filtre Topfiltre til flasker
The Stericup® vacuum filtration system is the most efficient single-use, disposable process for sterile media preparation. It combines a Steritop® filter unit with a receiver flask for processing and storing volumes from 150 to 1000 ml in a single, ready-to-use format. Its no-tip, easy-grip flask design and balanced profile improve stability during filtration in tight laminar flow hoods, and the convenient labeling and packaging make it easy to open and track each device.
  • Millipore Express® PLUS membrane for fastest flow rates and low protein binding to minimise absorption and protein loss, which affect downstream performance and costs
  • Durapore® membrane for ultra-low protein binding for minimum absorption and protein loss
  • Snug tight packaging, easy tear-open pouch, and convenient label designs make storing, opening and tracking your devices easy and efficient
  • Stable base and easy grip design of the Stericup® device enable any user to comfortably perform all stages of the workflow in a laminar flow hood
  • Recessed bottom of Stericup receiver flasks allows flasks to be stacked easily in refrigerated spaces
The bottom of the receiver flask is slightly recessed, so you can conveniently stack capped flasks after filtration, maximising precious refrigeration space. You can also use Steritop® bottle-top devices with your own sterile capture flask or bottle.
Applications:
Sterilisation of aqueous solutions (150 ml to 1 L); Sterile filtration of culture media and buffers with high-value protein or small molecule component.