PharmaDispo Disposable Powder Samplers
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5393-8000 5393-8001 5393-8010 5393-8011 5393-8020 5393-8021 5393-8030 5393-8031 5393-8040 5393-8041
BURK5393-8000EA 0 DKK
BURK5393-8000 BURK5393-8001 BURK5393-8010 BURK5393-8011 BURK5393-8020 BURK5393-8021 BURK5393-8030 BURK5393-8031 BURK5393-8040 BURK5393-8041
PharmaDispo Disposable Powder Samplers
Prøvetagere Prøvetagere til engangsbrug
PharmaDispo disposable powder sampler is a multi-target sampler that takes three separate target point samples with a single sampling.
  • Take small individual samples from different layers
  • Sterile versions are additionally gamma sterilised
  • Easy to use: Sliding mechanism with end stop for user-friendly operation
  • Hygienic and safe: Clean room manufactured, individually packaged, specifically designed for single use
The three separate sampling chambers of the PharmaDispo allow simultaneous sampling from different layers or zones. This allows the material composition to be investigated in a differentiated manner across multiple layers, which is particularly beneficial when a single sample does not sufficiently characterise the material. The target point samples taken can be analysed individually or combined to form a representative composite sample.
An advanced ergonomic design allows the sampler to sit comfortably in the hand and sampling can be performed with one hand. A sliding mechanism with an end stop makes it easy to open and close the sampler.
The PharmaDispo sampler accurately and safely takes small powder samples for quality control from open containers.
Individual packaging ensures that each sampler remains contamination free until used. Single-use eliminates the risk of cross-contamination between different samples or products. After a single use, the zone sampler is disposed of, ensuring the integrity of the work process.
Certificeringer: Manufactured, assembled and individually packaged under class 7 (ISO) clean room conditions.
Emballering: Samplers are supplied in a box of 20.