Showa XC500 Cut Resistant Gloves
Leverandør: Showa
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
XC500-6/S XC500-9/XL XC500-10/XXL XC500-7/M XC500-8/L
BESTXC500-6/SEA 1730 DKK
BESTXC500-6/S BESTXC500-9/XL BESTXC500-10/XXL BESTXC500-7/M BESTXC500-8/L
Showa XC500 Cut Resistant Gloves
Handsker
The new ultra-light XC500 combines high cut-resistance level E/A5 provided by a seamless HPPE/Stainless steel/Polyester liner with a durable, abrasive-resistant polyurethane palm coating usable in various industrial applications.
  • Ergonomic, flexible, lightweight and breathable
  • Increased dexterity
  • Seamless knit and form-fitting
  • Abrasion-resistant with rough grip
  • HPPE and stainless steel reinforced
  • Polyurethane palm coating
  • 12 Pair per polybag, 6 polybags per case, 72 pair per case
Trades and applications: Light assembly of oil-coated pieces, Maintenance, Mechanical and engineering, Repair glass and windows and Component assembly.
Certificeringer: EN 388:2016+A1:2018, ANSI/ISEA 105-2016 CUT.