Cut Resistant Gloves, XC800
Leverandør: Showa
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
XC800-10/XXL XC800-6/S XC800-7/M XC800-8/L XC800-9/XL
BESTXC800-10/XXLEA 2020 DKK
BESTXC800-10/XXL BESTXC800-6/S BESTXC800-7/M BESTXC800-8/L BESTXC800-9/XL
Cut Resistant Gloves, XC800
Handsker
The new ultra-light XC800 combines high cut-resistance level E/A5 provided by a seamless HPPE/stainless steel/pester liner with a durable, oil-resistant microporous nitrile palm coating usable in various industrial applications.
  • HPPE reinforced
  • Ergonomic
  • Microporous nitrile coating
  • Cut-resistant, abrasion-resistant
  • Flexible, breathable
  • Lightweight, form-fitting
  • Increased dexterity
  • Seamless knit
Application: Light assembly of oil-coated pieces, maintenance, mechanical and engineering, repair glass and windows, handling heavy equipment, intricate parts handling.
Certificeringer: ANSI/ISEA 105-2016 CUT, EN 388:2016.
Emballering: Pair per polybag: 12; Polybags per case: 6; Pair per case: 72.