Showa XC510 Cut-Resistant Industrial Gloves
Leverandør: Showa
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
XC510-6/S XC510-7/M XC510-8/L XC510-9/XL XC510-10/XXL
BESTXC510-6/SEA 1820 DKK
BESTXC510-6/S BESTXC510-7/M BESTXC510-8/L BESTXC510-9/XL BESTXC510-10/XXL
Showa XC510 Cut-Resistant Industrial Gloves
Handsker
The ultra-light XC510 industrial glove combines an 18-gauge UHMWPE cut-resistant liner with a durable, abrasive-resistant polyurethane palm coating for use in a wide variety of industrial applications.
  • Durable, form-fitting, flexible, and lightweight
  • Cut-resistance level D/A4
  • HPPE reinforced
  • Polyurethane palm coating
Flexible and breathable XC510 industrial gloves provide increased dexterity in a form-fitting, seamless knit design. These gloves are recommended for use in maintenance, glass/window repair, component assembly, engineering, and mechanical industries.
Certificeringer: Meets ANSI level A4. EN 388:2016+A1:2018, EN ISO 21420:2020. Cat. II.
Emballering: Gloves are packaged 12 pair per polybag, 6 polybags per case (72 total pairs).