Dimethoxy(diphenyl)silan ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

L04031.18 L04031.30 L04031.0B
L04031.18EA 412 DKK
L04031.18 L04031.30 L04031.0B
Dimethoxy(diphenyl)silan ≥97%
Dimethoxy(diphenyl)silan

Purity: 97% CAS No.: 6843-66-9 Molecular Formula: C14H16O2Si Formula Weight: 244.37 Flash Point: 121°(249°F) Refractive Index: 1.5420 Boiling Point: 160-162°/15mm Beilstein Reference: 2940458 MDL No.: MFCD00025690

Formel: C₁₄H₁₆O₂Si
Kogepunkt: 160…162 °C (15 mmHg)
Massefylde: 1,079
Flammepunkt: 121 °C (249 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00025690
CAS-nummer: 6843-66-9
EINECS: 229-929-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR