Pyridoxin hydrochlorid
Leverandør: Roth Carl
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Adermine hydrochloride, Vitamin B6 hydrochloride, PN HCl, Pyridoxine HCl, (5-Hydroxy-6-methylpyridine-3,4-diyl)dimethanol hydrochloride, Pyridoxol hydrochloride, Pyridoxine HCI
T914.1
T914.2
ROTHT914.1EA
272.44
DKK
ROTHT914.1
ROTHT914.2
Pyridoxin hydrochlorid
Pyridoxin hydrochlorid
Formel:
C₈H₁₂ClNO₃ Molekylvægt: 205.64 g/mol Smeltepunkt: 214…215 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00012807 CAS-nummer: 58-56-0 EINECS: 200-386-2 Merck Index: 13,08072 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...