L(-)-2-Amino-3-phenyl-1-propanol ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: (S)-(-)-2-Amino-3-phenyl-1-propanol, (S)-(-)-Phenylalaninol, (S)-2-Amino-3-phenylpropan-1-ol, L(-)-2-Amino-3-phenyl-1-propanol, L(-)-Phenylalaninol, H-Phenylalaninol, L(-)-2-Amino-3-phenylpropan-1-ol, (S)-(-)-2-Amino-3-phenylpropan-1-ol

A11586.03 A11586.06 A11586.14
A11586.03EA 341 DKK
A11586.03 A11586.06 A11586.14
L(-)-2-Amino-3-phenyl-1-propanol ≥98%
L(-)-2-Amino-3-phenyl-1-propanol

Purity: 98% CAS No.: 3182-95-4 Molecular Formula: C9H13NO Formula Weight: 151.21 Melting Point: 89-94° Beilstein Reference: 2208238 UN No.: 3259 MDL No.: MFCD00004732

Formel: C₉H₁₃NO
Molekylvægt: 151.21 g/mol
Kogepunkt: 278
Smeltepunkt: 89…94 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00004732
CAS-nummer: 3182-95-4
EINECS: 221-674-4
UN nummer: 3259
ADR: 8 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR