Isopropyl-β-D-thiogalactopyranoside ≥99% til molekylærbiologi
Leverandør: MP Biomedicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
0219402983 0219402990 0219402980 0219402991 0219402994 0219402995
ICNA0219402983EA 349 DKK
ICNA0219402983 ICNA0219402990 ICNA0219402980 ICNA0219402991 ICNA0219402994 ICNA0219402995
Isopropyl-β-D-thiogalactopyranoside ≥99% til molekylærbiologi
Isopropyl-β-D-thiogalactopyranoside
Cell culture media component for use in molecular genetics. β-Galactoside inducer.
Storage temperature: -20°C
pH: 5.0 - 7.0 (4% aq soln)
Formel: C₉H₁₈O₅S
Kogepunkt: 402 °C (1013 hPa)
Opbevaringstemperatur: Fryser
MDL Number: MFCD00063273
CAS-nummer: 367-93-1
EINECS: 206-703-0